Tisk…

(R)-(-)-Epichlorhydrin ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane

L14198.03 L14198.14 L14198.06
L14198.03EA 1850 CZK
L14198.03 L14198.14 L14198.06
(R)-(-)-Epichlorhydrin ≥98%
(R)-(-)-Epichlorhydrin

Purity: 98+% CAS No.: 51594-55-9 Molecular Formula: C3H5ClO Formula Weight: 92.53 Storage Temperature: 2°C to 8°C Flash Point: 32°(90°F) Refractive Index: 1.4380 Boiling Point: 116-117° Beilstein Reference: 1420785 Optical Rotation: -35° (c=1 in methanol) UN No.: 2023 MDL No.: MFCD00077759

Vzorec: ClCH₂CH(O)CH₂
MW: 92,52 g/mol
Bod varu: 116…117 °C
Bod tání: –48 °C
Hustota: 1,180
Bod vzplanutí: 32 °C (90 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00077759
CAS číslo: 51594-55-9
EINECS: 424-280-2
UN: 2023
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR