(±)-1,2-Propandiol ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
A18406.0E
A18406.36
A18406.0EEA
2280
CZK
A18406.0E
A18406.36
(±)-1,2-Propandiol ≥98%
(±)-1,2-Propandiol
Purity: 98+%
CAS No.: 57-55-6
Molecular Formula: C3H8O2
Formula Weight: 76.10
Flash Point: 107°(225°F)
Refractive Index: 1.4320
Odour: Nearly Odourless
pH: neutral
Melting Point: -60°
Boiling Point: 186-188°
Beilstein Reference: 1340498
Merck Reference: 14,7855
MDL No.: MFCD00064272
Vzorec:
CH₃CH(OH)CH₂OH MW: 76.1 Bod varu: 186…188 °C Bod tání: –60 °C Hustota: 1.036 Bod vzplanutí: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064272 CAS číslo: 57-55-6 EINECS: 200-338-0 Merck Index: 13,07947 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...