(±)-2-Butanol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
Purity: 99%
CAS No.: 78-92-2
Molecular Formula: CH3CH2CH(OH)CH3
Formula Weight: 74.12
Flash Point: 26°(79°F)
Refractive Index: 1.3970
Odour: Sweet
Melting Point: -115°
Boiling Point: 98-101°
Beilstein Reference: 1718765
Merck Reference: 14,1541
UN No.: 1120
MDL No.: MFCD00004569
Vzorec:
H₃CCH₂CH(OH)CH₃ Bod varu: 98…101 °C Bod tání: –115 °C Hustota: 0,808 Bod vzplanutí: 26 °C (79 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004569 CAS číslo: 78-92-2 EINECS: 201-158-5 UN: 1120 ADR: 3,III Merck Index: 14,1541 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...