(±)-Epichlorhydrin, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane
M-Clarity™ quality level = MQ200
45340-1L-F
45340-2.5L-F
45340-500ML-F
SIAL45340-1L-FEA
2050
CZK
SIAL45340-1L-F
SIAL45340-2.5L-F
SIAL45340-500ML-F
(±)-Epichlorhydrin, Sigma-Aldrich®
(±)-Epichlorhydrin
Vzorec:
C₃H₅ClO Bod varu: 117,9 °C (1013 hPa) Hustota: 0,9985 g/cm³ (20 °C) Bod vzplanutí: 28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005132 CAS číslo: 106-89-8 EINECS: 203-439-8 UN: 2023 ADR: 6.1,II Merck Index: 13,03642 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...