Tisk…

(±)-Epichlorhydrin 99%

Dodavatel: Thermo Scientific

Synonyms: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane

117780010 117780025 117780250 117780100
ACRO117780010EA 2450 CZK
ACRO117780010 ACRO117780025 ACRO117780250 ACRO117780100
(±)-Epichlorhydrin 99%
(±)-Epichlorhydrin
Vzorec: C₃H₅ClO
MW: 92,52 g/mol
MDL Number: MFCD00005132
CAS číslo: 106-89-8
Merck Index: 13,03642

Objednat

Výsledky specifikačního testu

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4365 to 1.4385 (20°C, 589 nm)
Water ≤0.02 % (K.F.)
GC ≥98.5 %
Color scale ≤15 APHA
Silicate and phosphate ≤5 ppm (as SiO2)
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm
Nitrogen compounds (as N) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR