Tisk…

Polyphosphoric acid ≥84% (as phosphorus pentoxide)

Dodavatel: Thermo Scientific
Nebezpečí

L14856.AE L14856.AP
L14856.AEEA 1100 CZK
L14856.AE L14856.AP
Polyphosphoric acid ≥84% (as phosphorus pentoxide)
Polyphosphoric acid

Purity: ca 84% CAS No.: 8017-16-1 Odour: Odourless Merck Reference: 14,7580 UN No.: 3264 MDL No.: MFCD00084480

Vzorec: H(PO₃H)nOH
Hustota: 2,1
Storage Temperature: Ambient
MDL Number: MFCD00084480
CAS číslo: 8017-16-1
EINECS: 232-417-0
UN: 3264
ADR: 8,III
Merck Index: 14,7580

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR