Tisk…
Vented caps for Falcon® flasks
Dodavatel: Corning
354638 354637 354639
734-1067EA 4230 CZK
734-1067 734-1066 734-1068
Vented caps for Falcon® flasks
Uzávěry Uzávěry k láhvím
PE with integral hydophobic 0,2 µm microporous membrane filter vent.
Vented caps minimise contamination associated with standard open incubation. Gases required for cell growth and metabolism pass freely through the vent, while microorganisms do not.