2-Methyl-2-pentenal ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 97%
CAS No.: 623-36-9
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 31°(87°F)
Refractive Index: 1.4495
Odour: Pleasant
Melting Point: -90°
Boiling Point: 134-136°
Beilstein Reference: 506124
UN No.: 1988
MDL No.: MFCD00006978
Vzorec:
C₆H₁₀O Bod varu: 134…136 °C Bod tání: –90 °C Hustota: 0,860 Bod vzplanutí: 31 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006978 CAS číslo: 623-36-9 EINECS: 210-789-5 UN: 1988 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...