Rhenium 99,99 Metals basis, powder -325 mesh
Dodavatel: Thermo Fisher Scientific
|
Varování
|
10313.09
10313.18
10313.04
10313.SS
10313.09EA
17090
CZK
10313.09
10313.18
10313.04
10313.SS
Rhenium 99,99 Metals basis, powder -325 mesh
Rhenium
Purity: 99.99%
CAS No.: 7440-15-5
Appearance: Black to grey
UN No.: 3089
MDL No.: MFCD00011195
Vzorec:
Re MW: 186.21 Bod varu: 5630 Bod tání: 3180 Hustota: 21.02 Storage Temperature: Ambient |
MDL Number:
MFCD00011195 CAS číslo: 7440-15-5 EINECS: 231-124-5 UN: 3089 ADR: 4.1 III Merck Index: 12,08343 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...