Tisk…

zinek ≥97%, powder -100 mesh

Dodavatel: Thermo Scientific
Nebezpečí

L13310.22 L13310.36 L13310.0E
L13310.22EA 944 CZK
L13310.22 L13310.36 L13310.0E
zinek ≥97%, powder -100 mesh
zinek

Purity: 97+% CAS No.: 7440-66-6 Physical Form: Powder Density: 7.13 Sensitivity: Air and Moisture Sensitive Appearance: Grey Odour: Odourless Melting Point: 420° Boiling Point: 907° UN No.: 1436 MDL No.: MFCD00011291

Vzorec: Zn
MW: 65,39 g/mol
Bod varu: 907 °C
Bod tání: 420 °C
Hustota: 7,13
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS číslo: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR