Tisk…

zinek ≥99.9% (metals basis), powder -140+325 mesh

Dodavatel: Thermo Fisher Scientific
Nebezpečí

39694.22 39694.36
39694.22EA 7550 CZK
39694.22 39694.36
zinek ≥99.9% (metals basis), powder -140+325 mesh
zinek

Purity: 99.9% CAS No.: 7440-66-6 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: Gray, bluish-white Odour: Odourless UN No.: 1436 MDL No.: MFCD00011291

Vzorec: Zn
Bod varu: 907 °C (1013 hPa)
Hustota: 7,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS číslo: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR