Tisk…

zinek, granule, EMSURE® ISO analytical reagent, Supelco®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ300

1.08780.0500 1.08780.1000
1.08780.0500EA 4130 CZK
1.08780.0500 1.08780.1000
zinek, granule, EMSURE® ISO analytical reagent, Supelco®
zinek

*1 AOX Enrichment Set  1.00678.0001 and AOX Sample Preparation Set 1.00677 required


*3 BOD Nutrient Salt Mixture  1.00687 required


*4 Reagents for free chlorine testing are Reagent Cl2-1 1.00086.0001  and Reagent Cl2-2 1.00087.0001 together with empty cell and accessories  1.00089.0001


Reagents for total chlorine are Reagent Cl2-1 1.00086.0001, Reagent Cl2-2 1.00087.0001 and Reagent Cl2-3 1.00088.0001  together with empty cell and accessories  1.00089.0001


*5 TOC screw caps for digestion required


*6 Zinc Reagent-6 required

Vzorec: Zn
MW: 65.39
Bod varu: 908
Bod tání: 420
Hustota: 7.14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS číslo: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9 III
Merck Index: 12,10255

Objednat

Výsledky specifikačního testu

Solubility (20 °C) insoluble
Melting Point 419 °C
Molar Mass 65.39 g/mol
Boiling Point 906 °C (1013 hPa)
Density 7.13 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR