Tisk…

zinek ≥99.95% (metals basis), wire, Ø 1.0 mm (0.04 in)

Dodavatel: Thermo Fisher Scientific

42705.G1 42705.G5 42705.G9
42705.G1EA 1780 CZK
42705.G1 42705.G5 42705.G9
zinek ≥99.95% (metals basis), wire, Ø 1.0 mm (0.04 in)
zinek

Purity: 99.95% CAS No.: 7440-66-6 Physical Form: ≈5.60g/m Odour: Odourless MDL No.: MFCD00011291

Vzorec: Zn
MW: 65.39
Bod varu: 908
Bod tání: 420
Hustota: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS číslo: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR