zinek ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Dodavatel: Thermo Fisher Scientific
42703.G5
42703.G9
42703.G5EA
4090
CZK
42703.G5
42703.G9
zinek ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
zinek
Purity: 99.95%
CAS No.: 7440-66-6
Physical Form: ≈0.35g/m
Odour: Odourless
MDL No.: MFCD00011291
Vzorec:
Zn MW: 65.39 Bod varu: 908 Bod tání: 420 Hustota: 7.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS číslo: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...