Tisk…

zinek ≥99.99% (metals basis), shot, Ø 10mm (0.4in), Thickness: 2 mm (0.08 in)

Dodavatel: Thermo Fisher Scientific

10440.36 10440.A3 10440.22
10440.36EA 3150 CZK
10440.36 10440.A3 10440.22
zinek ≥99.99% (metals basis), shot, Ø 10mm (0.4in), Thickness: 2 mm (0.08 in)
zinek

Purity: 99.99% CAS No.: 7440-66-6 Odour: Odourless MDL No.: MFCD00011291

Vzorec: Zn
Bod varu: 908 °C (1013 hPa)
Bod tání: 420 °C
Hustota: 7,14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS číslo: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR