zinek ≥99.99% (metals basis), ingot, Length: 345 mm (13.6 in)
Dodavatel: Thermo Fisher Scientific
00682.KF
00682.KJ
00682.KFEA
1740
CZK
00682.KF
00682.KJ
zinek ≥99.99% (metals basis), ingot, Length: 345 mm (13.6 in)
zinek
Purity: 99.99%
CAS No.: 7440-66-6
Physical Form: ≈175g/ingot
Odour: Odourless
MDL No.: MFCD00011291
Vzorec:
Zn MW: 65.39 Bod varu: 908 Bod tání: 420 Hustota: 7.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS číslo: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...