Tisk…

měď ≥99.999% (metals basis), foil, Puratronic®, Thickness: 0.025 mm (0.001 in)

Dodavatel: Thermo Fisher Scientific

10950.GZ 10950.HV
10950.GZEA 6660 CZK
10950.GZ 10950.HV
měď ≥99.999% (metals basis), foil, Puratronic®, Thickness: 0.025 mm (0.001 in)
měď

Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈1.1g/25x200mm Odour: Odourless MDL No.: MFCD00010965

Vzorec: Cu
MW: 63.55
Bod varu: 2595
Bod tání: 1083
Hustota: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS číslo: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR