Tisk…

Praseodymium ≥99.5% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)

Dodavatel: Thermo Scientific

13963.FF 13963.FI
13963.FFEA 7180 CZK
13963.FF 13963.FI
Praseodymium ≥99.5% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Praseodymium

Purity: 99.5% CAS No.: 7440-10-0 Physical Form: ≈4.23g/25x25mm, Packaged in mylar under argon Appearance: White Odour: Odourless MDL No.: MFCD00011174

Vzorec: Pr
Storage Temperature: Ambient
MDL Number: MFCD00011174
CAS číslo: 7440-10-0
EINECS: 231-120-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR