Tisk…

Lantan ≥99.9% (REO, rare earth oxide basis), rods, Ø 6.35 mm (0.25 in)

Dodavatel: Thermo Scientific
Nebezpečí

40292.DH 40292.DJ 40292.DM 40292.HF
40292.DHEA 7070 CZK
40292.DH 40292.DJ 40292.DM 40292.HF
Lantan ≥99.9% (REO, rare earth oxide basis), rods, Ø 6.35 mm (0.25 in)
Lantan

Purity: 99.9% CAS No.: 7439-91-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.20g/mm, Cast rod, diameter is approximate Appearance: Gray to silver UN No.: 3208 MDL No.: MFCD00011066

Vzorec: La
MW: 138,91 g/mol
Bod varu: 3464 °C (1013 hPa)
Bod tání: 920 °C
Hustota: ∼6,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS číslo: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR