Tisk…

Telur ≥99.99% (metals basis), powder -325 mesh

Dodavatel: Thermo Fisher Scientific
Nebezpečí

43450.09 43450.18
43450.09EA 2140 CZK
43450.09 43450.18
Telur ≥99.99% (metals basis), powder -325 mesh
Telur

Purity: 99.99% CAS No.: 13494-80-9 Odour: Odourless UN No.: 3288 MDL No.: MFCD00134062

Vzorec: Te
MW: 127.6
Bod varu: 990
Bod tání: 450
Hustota: 8.83
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS číslo: 13494-80-9
EINECS: 236-813-4
UN: 3288
ADR: 6.1 III
Merck Index: 12,09273

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR