Tisk…

Telur ≥99.9999% (metals basis), pieces, zone refined

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11076.18 11076.30 11076.09
11076.18EA 9750 CZK
11076.18 11076.30 11076.09
Telur ≥99.9999% (metals basis), pieces, zone refined
Telur

Purity: 99.9999% CAS No.: 13494-80-9 Odour: Odourless MDL No.: MFCD00134062

Vzorec: Te
MW: 127.6
Bod varu: 990
Bod tání: 450
Hustota: 8.83
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS číslo: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1 III
Merck Index: 12,09273

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR