Telur ≥99.5% (metals basis), powder -200 mesh
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
10605.18
10605.30
10605.18EA
3160
CZK
10605.18
10605.30
Telur ≥99.5% (metals basis), powder -200 mesh
Telur
Purity: 99.5%
CAS No.: 13494-80-9
Appearance: Grey
Odour: Odourless
UN No.: 3288
MDL No.: MFCD00134062
Vzorec:
Te MW: 127.6 Bod varu: 990 Bod tání: 450 Hustota: 8.83 Storage Temperature: Ambient |
MDL Number:
MFCD00134062 CAS číslo: 13494-80-9 EINECS: 236-813-4 UN: 3288 ADR: 6.1 III Merck Index: 12,09273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...