Tisk…

Olovo ≥99.995% (metals basis), wire, Ø 0.75 mm (0.03 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

44809.G2 44809.G6
44809.G2EA 3810 CZK
44809.G2 44809.G6
Olovo ≥99.995% (metals basis), wire, Ø 0.75 mm (0.03 in)
Olovo

Purity: 99.995% CAS No.: 7439-92-1 Physical Form: ≈5g/m Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Vzorec: Pb
MW: 207.2
Bod varu: 1740
Bod tání: 327
Hustota: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS číslo: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR