Tisk…

Olovo ≥99.999% (metals basis), rods, Ø 6.35 mm (0.25 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

42691.DJ 42691.DT
42691.DJEA 2510 CZK
42691.DJ 42691.DT
Olovo ≥99.999% (metals basis), rods, Ø 6.35 mm (0.25 in)
Olovo

Purity: 99.999% CAS No.: 7439-92-1 Physical Form: ≈18g/50mm Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Vzorec: Pb
Bod varu: 1740 °C (1013 hPa)
Bod tání: 327 °C
Hustota: 11,3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS číslo: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR