Tisk…

Olovo ≥99.9% (metals basis), plate, Thickness: 6.35 mm (0.25 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

12603.RF 12603.VA
12603.RFEA 4770 CZK
12603.RF 12603.VA
Olovo ≥99.9% (metals basis), plate, Thickness: 6.35 mm (0.25 in)
Olovo

Purity: 99.9% CAS No.: 7439-92-1 Physical Form: ≈1.6kg/15x15cm Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Vzorec: Pb
Bod varu: 1740 °C (1013 hPa)
Bod tání: 327 °C
Hustota: 11,3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS číslo: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR