Tisk…
Culture dishes, Corning® BioCoat™ Matrigel® matrix
Dodavatel: Corning
354601 354600
734-0232EA 7790 CZK
734-0232 734-0231
Culture dishes, Corning® BioCoat™ Matrigel® matrix
Misky kultivační Misky pro tkáňové kultury
Corning Matrigel basement membrane matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in ECM proteins. Its major component is laminin, followed by collagen IV, heparan sulfate proteoglycans, entactin, and nidogen.
Corning Matrigel matrix is effective for the attachment and differentiation of both normal and transformed anchorage-dependent epithelial and other cell types including neurons and oligodendrocytes.