Tisk…

Olovo ≥99.9998% (metals basis), rods, Puratronic®, Ø 12.7 mm (0.5 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11346.18 11346.97 11346.98
11346.18EA 3100 CZK
11346.18 11346.97 11346.98
Olovo ≥99.9998% (metals basis), rods, Puratronic®, Ø 12.7 mm (0.5 in)
Olovo

Purity: 99.9998% CAS No.: 7439-92-1 Physical Form: ≈3.5cm/50g, may be in multiple pieces Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Vzorec: Pb
Bod varu: 1740 °C (1013 hPa)
Bod tání: 327 °C
Hustota: 11,3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS číslo: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR