Tisk…

Olovo ≥99.9% (metals basis), powder -200 mesh

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10223.30 10223.A1 10223.A7
10223.30EA 1850 CZK
10223.30 10223.A1 10223.A7
Olovo ≥99.9% (metals basis), powder -200 mesh
Olovo

Purity: 99.9% CAS No.: 7439-92-1 Odour: Odourless UN No.: 3077 MDL No.: MFCD00134050

Vzorec: Pb
Bod varu: 1749 °C (1013 hPa)
Hustota: 11,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS číslo: 7439-92-1
EINECS: 231-100-4
UN: 3077
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR