Tisk…

Olovo ≥97% (metals basis), shot 4 mm (0.2 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

00675.36 00675.A3 00675.22
00675.36EA 2860 CZK
00675.36 00675.A3 00675.22
Olovo ≥97% (metals basis), shot 4 mm (0.2 in)
Olovo

Purity: 97% CAS No.: 7439-92-1 Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Vzorec: Pb
Bod varu: 1740 °C (1013 hPa)
Bod tání: 327 °C
Hustota: 11,3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS číslo: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR