Tisk…

Sodík ≥99.8% (metals basis), ingot

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10342.35 10342.60
10342.35EA 4880 CZK
10342.35 10342.60
Sodík ≥99.8% (metals basis), ingot
Sodík

Purity: 99.8% CAS No.: 7440-23-5 Appearance: Silver-white Odour: Odourless UN No.: 1428 MDL No.: MFCD00085307

Vzorec: Na
Bod varu: 881 °C (1013 hPa)
Hustota: 0,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085307
CAS číslo: 7440-23-5
EINECS: 231-132-9
UN: 1428
ADR: 4.3,I
Merck Index: 13,08641

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR