Sodík ≥99.95% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Breakseal ampoule
10336.03
10336.06
10336.03EA
5260
CZK
10336.03
10336.06
Sodík ≥99.95% (metals basis)
Sodík
Purity: 99.95%
CAS No.: 7440-23-5
Physical Form: Sealed in breakseal ampoules
Odour: Odourless
UN No.: 1428
MDL No.: MFCD00085307
Vzorec:
Na MW: 22.99 Bod varu: 881 Bod tání: 97.8 Hustota: 0.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085307 CAS číslo: 7440-23-5 EINECS: 231-132-9 UN: 1428 ADR: 4.3 I Merck Index: 13,08641 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...