Draslík ≥99.95% (metals basis), solid
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Ampuled under argon
10299.06
10299.03
10299.06EA
12500
CZK
10299.06
10299.03
Draslík ≥99.95% (metals basis), solid
Draslík
Purity: 99.95%
CAS No.: 7440-09-7
Physical Form: Sealed in standard ampoules under vacuum
Odour: Odourless
UN No.: 2257
MDL No.: MFCD00133776
Vzorec:
K MW: 39.1 Bod varu: 760 Bod tání: 63 Hustota: 0.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133776 CAS číslo: 7440-09-7 EINECS: 231-119-8 UN: 2257 ADR: 4.3 I Merck Index: 12,07763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...