Cesium ≥99.98% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Breakseal ampoule
10146.03
10146.06
10146.14
10146.03EA
5360
CZK
10146.03
10146.06
10146.14
Cesium ≥99.98% (metals basis)
Cesium
Purity: 99.98%
CAS No.: 7440-46-2
Physical Form: Vacuum sealed in breakseal ampoules
Appearance: Golden
Odour: Odourless
UN No.: 1407
MDL No.: MFCD00134037
Vzorec:
Cs Bod varu: 705 °C (1013 hPa) Hustota: 1,873 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134037 CAS číslo: 7440-46-2 EINECS: 231-155-4 UN: 1407 ADR: 4.3,I Merck Index: 12,02051 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...