Tisk…
Cell culture dishes, amine and carboxyl coated, Corning® PureCoat™
Dodavatel: Corning
354732 354784 356784 356732
734-1481EA 2200 CZK
734-1481 734-1489 734-1490 734-1482
Cell culture dishes, amine and carboxyl coated, Corning® PureCoat™
Misky kultivační Misky pro tkáňové kultury
PS. Corning® PureCoat™ surfaces are a novel family of chemically-defined, animal-free cell culture surfaces designed to enhance cell performance. A proprietary thin-film coating technology produces a uniform, functionalised surface that provides a highly controlled environment for cell culture applications.
  • Available as amine (positively charged) or carboxyl (negatively charged) surface chemistries
  • Optimised for culture in low serum or no serum conditions
  • Enhanced freeze-thaw recovery of cryopreserved cells
  • Improved differentiation of cells
  • Culture of many cell lines and primary cells supported, including primary neuronal cells and stem cells
A proprietary thin-film coating technology produces a uniform, functionalized surface that provides a highly controlled environment for cell culture applications.