Tisk…
Urine sample kits, Nunc™
Dodavatel: Thermo Fisher Scientific
Nunc™
391-7786EA 13980 CZK
391-7786
Urine sample kits, Nunc™
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Transport and universal containers comply with 95 kPascal test described in UN Packaging Instructions 602/650 for transport of infectious substances and diagnostic specimens.
  • Sterile, Easy to use
  • Can be used by the patient at home
  • EAN barcode on the bag of product (Cat. No. 391-7783)
  • CE-marked