Tisk…

Capsaicin, Sigma-Aldrich®

Dodavatel: Merck

21750-100MG-F 21750-1G-F
SIAL21750-100MG-FEA 2060 CZK
SIAL21750-100MG-F SIAL21750-1G-F
Capsaicin, Sigma-Aldrich®
Capsaicin
Vzorec: C₁₈H₂₇NO₃
Bod varu: 210…220 °C (0,01 mmHg)
MDL Number: MFCD00017259
CAS číslo: 404-86-4
EINECS: 206-969-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,01811

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR