Tisk…
Síran měďnatý 0.5 mol/l Reag. Ph. Eur. 1022501
Dodavatel: VWR Chemicals
Varování
87808.290EA 4194 CZK
87808.290
Síran měďnatý 0.5 mol/l Reag. Ph. Eur. 1022501
Síran měďnatý
Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Vzorec: CuSO₄
MW: 159,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS číslo: 7758-98-7
EINECS: 231-847-6
UN: 3082
ADR: 9,III
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022
Objednat
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022