CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid
M-Clarity™ quality level = MQ200
C2278-100G
C2278-1KG
C2278-25G
C2278-500G
SIALC2278-100GEA
8150
CZK
SIALC2278-100G
SIALC2278-1KG
SIALC2278-25G
SIALC2278-500G
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid), Sigma-Aldrich®
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid)
Vzorec:
C₉H₁₉NO₄S MW: 237,32 g/mol Bod tání: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041778 CAS číslo: 73463-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...