Tisk…

Phthalocyanine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Pigment Blue 16, 29H,31H-Phthalocyanine

253103-1G 253103-5G
SIAL253103-1GEA 1420 CZK
SIAL253103-1G SIAL253103-5G
Phthalocyanine, Sigma-Aldrich®
Phthalocyanine
Vzorec: C₃₂H₁₈N₈
MW: 514.55
Bod tání: 210
Hustota: 1.447 g/cm³ (–113 °C)
MDL Number: MFCD00005085
CAS číslo: 574-93-6
EINECS: 209-378-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR