Tisk…

Phthalocyanine

Dodavatel: Thermo Fisher Scientific
Varování

32073.03 32073.06 32073.14
32073.03EA 1160 CZK
32073.03 32073.06 32073.14
Phthalocyanine
Phthalocyanine

CAS No.: 574-93-6 Molecular Formula: C32H18N8 Formula Weight: 514.52 Physical Form: Crystalline Odour: Odourless MDL No.: MFCD00005085

Vzorec: C₃₂H₁₈N₈
Bod tání: 210 °C
Hustota: 1,447 g/cm³ (–113 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005085
CAS číslo: 574-93-6
EINECS: 209-378-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR