Tisk…

di-Sodium β-glycerophosphate pentahydrate, Millipore®

Dodavatel: Merck Millipore Calbiochem

Synonyms: Sodium β-glycerophosphate pentahydrate, Disodium-β-glycerophosphate pentahydrate, Glycerol-2-phosphate, β-GP, di-Sodium-β-glycerophosphate pentahydrate, β-Glycerophosphoric acid disodium salt pentahydrate, β-glycerolphosphate

35675-100 35675-50
35675-100EA 4340 CZK
35675-100 35675-50
di-Sodium β-glycerophosphate pentahydrate, Millipore®
di-Sodium β-glycerophosphate pentahydrate
Vzorec: C₃H₇Na₂O₆P·5H₂O
MDL Number: MFCD00002135
CAS číslo: 13408-09-8
EINECS: 212-464-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR