beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
NADP sodium hydrate, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium monosodium salt xhydrate
MQ200
N3139-100MG
N3139-1G
N3139-250MG
SIALN3139-100MGEA
3590
CZK
SIALN3139-100MG
SIALN3139-1G
SIALN3139-250MG
beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate, Sigma-Aldrich®
beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate
Vzorec:
C₂₁H₂₇N₇NaO₁₇P₃·xH₂O |
MDL Number:
MFCD00150480 CAS číslo: 698999-85-8 EINECS: 214-664-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...