Copper(II) phthalocyanine 90% (dye content) β-form, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15
M-Clarity™ quality level = MQ100
252980-25G
252980-100G
252980-5G
SIAL252980-25GEA
2460
CZK
SIAL252980-25G
SIAL252980-100G
SIAL252980-5G
Copper(II) phthalocyanine 90% (dye content) β-form, Sigma-Aldrich®
Copper(II) phthalocyanine
Vzorec:
C₃₂H₁₆CuN₈ MW: 576,08 g/mol Bod tání: 480 °C |
MDL Number:
MFCD00010719 CAS číslo: 147-14-8 EINECS: 205-685-1 Merck Index: 12,02584 |
Výsledky specifikačního testu
Appearance (Colour) | Dark Blue to Violet or Purple |
Appearance (Form) | Powder |
Infrared Spectrum | Conforms to Structure |
ICP Major Analysis | Confirmed |
Wavelength, c=0.005 g/l; Chloronapthalene | 675 - 681 nm |
Extinction Coefficient | ≥75000 |
Carbon | 58.7 - 74.7% |
Nitrogen | 17.1 - 21.7% |
Solubility (Turbidity) | Turbid |
Solubility (Color) c = 10 mg/ml; Pyridine | Dark Blue |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...