Copper(II) phthalocyanine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15
M-Clarity™ quality level = MQ100
546682-200MG
546682-2G
SIAL546682-200MGEA
0
CZK
SIAL546682-200MG
SIAL546682-2G
Copper(II) phthalocyanine, Sigma-Aldrich®
Copper(II) phthalocyanine
Vzorec:
C₃₂H₁₆CuN₈ MW: 576,08 g/mol Bod tání: 480 °C |
MDL Number:
MFCD00010719 CAS číslo: 147-14-8 EINECS: 205-685-1 Merck Index: 12,02584 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...