Tisk…

Copper(II) phthalocyanine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15

M-Clarity™ quality level = MQ100

546674-1G 546674-5G
SIAL546674-1GEA 11720 CZK
SIAL546674-1G SIAL546674-5G
Copper(II) phthalocyanine, Sigma-Aldrich®
Copper(II) phthalocyanine
Vzorec: C₃₂H₁₆CuN₈
MW: 576.08
Bod tání: 480
MDL Number: MFCD00010719
CAS číslo: 147-14-8
EINECS: 205-685-1
Merck Index: 12,02584

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR