Amberlite® IRN-77, ion exchange resin cationic type (H⁺) nuclear grade
Dodavatel: Thermo Scientific
16821.36
16821.A3
16821.36EA
2240
CZK
16821.36
16821.A3
Amberlite® IRN-77, ion exchange resin cationic type (H⁺) nuclear grade
Amberlite® IRN-77, ion exchange resin cationic type (H⁺)
CAS No.: 11128-94-2
Molecular Formula: Styrene-DVB; Strongly acidic cation exchange resin
Merck Reference: 14,382
MDL No.: MFCD00145822
Storage Temperature:
Ambient |
CAS číslo:
11128-94-2 Merck Index: 14,382 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...