Tisk…

Sodium dodecylbenzenesulphonate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Laurylbenzenesulphonic acid Sodium Salt, Alkylbenzenesulphonic acid Sodium Salt, Sodium Alkylbenzenesulphonate, Dodecylbenzenesulphonic acid sodium salt, Sodium Laurylbenzenesulphonate

M-Clarity™ quality level = MQ200

289957-500G 289957-25G 289957-10KG 289957-1KG
SIAL289957-500GEA 3090 CZK
SIAL289957-500G SIAL289957-25G SIAL289957-10KG SIAL289957-1KG
Sodium dodecylbenzenesulphonate, Sigma-Aldrich®
Sodium dodecylbenzenesulphonate
Vzorec: C₁₈H₂₉NaO₃S
Bod tání: > 300 °C
Hustota: 1 g/cm³ (20 °C)
MDL Number: MFCD00011508
CAS číslo: 25155-30-0
EINECS: 246-680-4
Merck Index: 13,08686

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR