Sodium dodecylbenzenesulphonate
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Laurylbenzenesulphonic acid Sodium Salt, Alkylbenzenesulphonic acid Sodium Salt, Sodium Alkylbenzenesulphonate, Dodecylbenzenesulphonic acid sodium salt, Sodium Laurylbenzenesulphonate
325910010
325910050
325910250
ACRO325910010EA
1690
CZK
ACRO325910010
ACRO325910050
ACRO325910250
Sodium dodecylbenzenesulphonate
Sodium dodecylbenzenesulphonate
Vzorec:
C₁₈H₂₉NaO₃S MW: 348,48 g/mol Bod tání: > 300 °C Hustota: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00011508 CAS číslo: 25155-30-0 EINECS: 246-680-4 Merck Index: 13,08686 |
Výsledky specifikačního testu
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥83 % |
Water | ≤2 % |
pH | 6.5 to 9.5 (1% soln.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...