Antimony (III) ethanolate ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
43097.22
43097.14
43097.06
43097.22EA
31610
CZK
43097.22
43097.14
43097.06
Antimony (III) ethanolate ≥99.9% (metals basis)
Antimony (III) ethanolate
Purity: 99.9%
CAS No.: 10433-06-4
Molecular Formula: Sb(OC2H5)3
Formula Weight: 256.90
Flash Point:
Vzorec:
C₆H₁₅O₃Sb MW: 256.94 Bod varu: 95 °C (11 mmHg) Hustota: 1.524 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015123 CAS číslo: 10433-06-4 EINECS: 233-914-5 UN: 3082 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...