Tisk…

9-Fluorenone-4-carboxylic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 9-Oxofluoren-4-carboxylic acid, 9-Oxo-9H-fluorene-4-carboxylic acid, 9-Oxofluorene-4-carboxylic acid

B25021.03 B25021.06
B25021.03EA 1010 CZK
B25021.03 B25021.06
9-Fluorenone-4-carboxylic acid ≥98%
9-Fluorenone-4-carboxylic acid

Purity: 98+% CAS No.: 6223-83-2 Molecular Formula: C14H8O3 Formula Weight: 224.22 Melting Point: 224-228° Beilstein Reference: 2694801

Vzorec: C₁₄H₈O₃
MW: 224,22 g/mol
Bod tání: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00001145
CAS číslo: 6223-83-2
EINECS: 228-311-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR